Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,7-difluorobenzo[d]thiazole 95%

Supplier: AMBEED, INC
Danger

A523253-100MG A523253-1G A523253-250MG
76830-378EA 60 USD InStock
76830-378 76830-380 76830-382
2-Chloro-4,7-difluorobenzo[d]thiazole 95%
2-Chloro-4,7-difluorobenzo[d]thiazole
Precaución: for Research Use Only


Formula: C₇H₂ClF₂NS
MDL Number: MFCD09748623
CAS Number: 1177320-67-0
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty