Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5,6-Tetrabromophenolsulfonephthalein 95%

Supplier: AMBEED, INC
Warning

A524233-25G A524233-5G
76830-384EA 556 USD InStock
76830-384 76830-386
3,4,5,6-Tetrabromophenolsulfonephthalein 95%
3,4,5,6-Tetrabromophenolsulfonephthalein
Precaución: for Research Use Only


Formula: C₁₉HBr₄O₅S
MW: 669.96 g/mol
MDL Number: MFCD00009744
CAS Number: 77172-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty