Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S,5S)-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate 96%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A528413-100MG A528413-1G A528413-250MG A528413-5G
76830-398EA 41 USD InStock
76830-398 76830-400 76830-402 77684-367
(3S,5S)-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate 96%, Ambeed.Inc
(3S,5S)-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate
Precaución: For research use only.


Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD08686694
CAS Number: 888327-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty