Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-chloro-2-methoxybenzoate 98%

Supplier: AMBEED, INC
Warning

A536102-1G A536102-250MG A536102-5G
76830-458EA 23 USD InStock
76830-458 76830-460 76830-462
Methyl-4-chloro-2-methoxybenzoate 98%
Methyl-4-chloro-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₉ClO₃
MDL Number: MFCD09800922
CAS Number: 78955-90-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty