Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5,6-diaminonicotinate 95%

Supplier: AMBEED, INC
Warning

A538843-100MG A538843-1G A538843-250MG
76830-474EA 25 USD InStock
76830-474 76830-476 76830-478
Methyl 5,6-diaminonicotinate 95%
Methyl 5,6-diaminonicotinate
Precaución: for Research Use Only


Formula: C₇H₉N₃O₂
MDL Number: MFCD11656314
CAS Number: 104685-76-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty