Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-amino-5-bromothiophene-2-carboxylate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A540640-100MG A540640-1G A540640-5G
76830-496EA 33 USD InStock
76830-496 77684-382 77684-383
Methyl 4-amino-5-bromothiophene-2-carboxylate 95%, Ambeed.Inc
Methyl 4-amino-5-bromothiophene-2-carboxylate
Precaución: For research use only.


Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
MDL Number: MFCD23135739
CAS Number: 89499-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty