Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl(2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl)silane 98%

Supplier: AMBEED, INC

A541253-1G A541253-250MG
76830-504EA 65 USD InStock
76830-504 76830-506
Trimethyl(2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl)silane 98%
Trimethyl(2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl)silane
Precaución: for Research Use Only


Formula: C₁₂H₂₂Si
MW: 194.38 g/mol
MDL Number: MFCD00674076
CAS Number: 134695-74-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty