Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%

Supplier: AMBEED, INC
Warning

A556440-100MG A556440-1G A556440-250MG
76830-588EA 35 USD InStock
76830-588 76830-590 76830-592
(S)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%
(S)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Precaución: for Research Use Only


Formula: C₃₆H₂₉O₄P
MDL Number: MFCD27952571
CAS Number: 1170736-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty