Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-Amino-2-fluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A557277-100MG A557277-1G A557277-250MG A557277-5G
76830-602EA 7 USD InStock
76830-602 76830-604 76830-606 76830-608
Methyl 3-Amino-2-fluorobenzoate 98%
Methyl 3-Amino-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₈FNO₂
MDL Number: MFCD12923265
CAS Number: 1195768-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty