Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)-4-bromoanisole 96%

Supplier: AMBEED, INC
Warning

A557784-1G A557784-250MG A557784-5G
76830-610EA 20 USD InStock
76830-610 76830-612 76830-614
2-(Benzyloxy)-4-bromoanisole 96%
2-(Benzyloxy)-4-bromoanisole
Precaución: for Research Use Only


Formula: C₁₄H₁₃BrO₂
MDL Number: MFCD09836381
CAS Number: 78504-28-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty