Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Isopentylsulphonyl)phenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A567677-100MG A567677-1G A567677-250MG
76830-692EA 45 USD InStock
76830-692 76830-694 76830-696
4-(Isopentylsulphonyl)phenylboronic acid 95%
4-(Isopentylsulphonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇BO₄S
MDL Number: MFCD12546540
CAS Number: 1217500-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty