Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-amino-4-((tert-butoxycarbonyl)amino)benzoate 95%

Supplier: AMBEED, INC
Warning

A577480-100MG A577480-250MG
76830-750EA 58 USD InStock
76830-750 76830-752
Methyl 2-amino-4-((tert-butoxycarbonyl)amino)benzoate 95%
Methyl 2-amino-4-((tert-butoxycarbonyl)amino)benzoate
Precaución: for Research Use Only


Formula: C₁₃H₁₈N₂O₄
MDL Number: MFCD13176796
CAS Number: 849792-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty