Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-2-(Propan-2-yl)morpholine 98%

Supplier: AMBEED, INC
Danger

A593807-100MG A593807-1G A593807-250MG A593807-5G
76830-860EA 34 USD InStock
76830-860 76830-862 76830-864 76830-866
(2S)-2-(Propan-2-yl)morpholine 98%
(2S)-2-(Propan-2-yl)morpholine
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD14582704
CAS Number: 1286768-31-7
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty