Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethoxy-1,3,5-triazin-2(1H)-one 95%

Supplier: AMBEED, INC
Warning

A598212-10G A598212-1G A598212-250MG
76830-890EA 172 USD InStock
76830-890 76830-892 76830-894
4,6-Dimethoxy-1,3,5-triazin-2(1H)-one 95%
4,6-Dimethoxy-1,3,5-triazin-2(1H)-one
Precaución: for Research Use Only


Formula: C₅H₇N₃O₃
MW: 157.127 g/mol
MDL Number: MFCD02932731
CAS Number: 1075-59-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty