Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Piperidine-1-carbonyloxy)phenylboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A606544-1G A606544-250MG A606544-5G
76830-962EA 163 USD InStock
76830-962 76830-964 76830-966
4-(Piperidine-1-carbonyloxy)phenylboronic acid pinacol ester 96%
4-(Piperidine-1-carbonyloxy)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₈H₂₆BNO₄
MW: 331.21 g/mol
MDL Number: MFCD09027287
CAS Number: 913836-28-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty