Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Propionyl)benzeneboronic acid 95%

Supplier: AMBEED, INC
Warning

A618672-100MG A618672-1G A618672-250MG A618672-5G
76831-078EA 36 USD InStock
76831-078 76831-080 76831-082 76831-084
4-(Propionyl)benzeneboronic acid 95%
4-(Propionyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BO₃
MDL Number: MFCD04039038
CAS Number: 186498-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty