Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluormethylthio)phenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A642908-100MG A642908-1G A642908-250MG
76831-240EA 15 USD InStock
76831-240 76831-242 76831-244
4-(Trifluormethylthio)phenylboronic acid pinacol ester 97%
4-(Trifluormethylthio)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₆BF₃O₂S
MW: 304.1400 g/mol
MDL Number: MFCD11042448
CAS Number: 1005206-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty