Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-bromo-4-fluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A650255-100G A650255-10G A650255-1G A650255-25G A650255-5G
76831-284EA 162 USD InStock
76831-284 76831-286 76831-288 76831-290 76831-292
Ethyl-3-bromo-4-fluorobenzoate 97%
Ethyl-3-bromo-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrFO₂
MDL Number: MFCD06204378
CAS Number: 23233-33-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty