Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2-chloropyridin-3-ylcarbamate 98%

Supplier: AMBEED, INC
Warning

A657869-100MG A657869-1G A657869-250MG A657869-5G
76831-388EA 6 USD InStock
76831-388 76831-390 76831-392 76831-394
tert-Butyl-2-chloropyridin-3-ylcarbamate 98%
tert-Butyl-2-chloropyridin-3-ylcarbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD12026344
CAS Number: 209798-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty