Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylenepiperidine hydrochloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A660956-25G A660956-5G A660956-1G A660956-100G
76831-406EA 42 USD InStock
76831-406 76831-408 77684-554 77684-553
4-Methylenepiperidine hydrochloride 98%, Ambeed.Inc
4-Methylenepiperidine hydrochloride
Precaución: For research use only.


Formula: C₆H₁₂ClN
MDL Number: MFCD17977038
CAS Number: 144230-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty