Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A668312-100MG A668312-250MG A668312-5G
76831-454EA 58 USD InStock
76831-454 76831-456 76831-458
Ethyl 5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylate hydrochloride 95%
Ethyl 5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD10566314
CAS Number: 1207175-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty