Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Methylvaleric acid 97%

Supplier: AMBEED, INC
Danger

A689144-100G A689144-10G
76831-544EA 54 USD InStock
76831-544 76831-546
(±)-2-Methylvaleric acid 97%
(±)-2-Methylvaleric acid
Precaución: for Research Use Only


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 195
Melting Pt: -85
Flash Pt: 87
MDL Number: MFCD00002671
CAS Number: 97-61-0
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty