Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Isobutoxy-5-methylpyridin-3-yl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A698878-1G A698878-250MG A698878-5G
76831-620EA 119 USD InStock
76831-620 76831-622 76831-624
(6-Isobutoxy-5-methylpyridin-3-yl)boronic acid 98%
(6-Isobutoxy-5-methylpyridin-3-yl)boronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₆BNO₃
MW: 209.05 g/mol
MDL Number: MFCD18087688
CAS Number: 1256355-19-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty