Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)pyrrolidine 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A712055-100MG A712055-250MG A712055-1G
76831-704EA 61 USD InStock
76831-704 76831-706 77684-629
1-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)pyrrolidine 98%, Ambeed.Inc
1-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)pyrrolidine
Precaución: For research use only.


Formula: C₁₈H₂₈BNO₃
MDL Number: MFCD09746211
CAS Number: 656257-46-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty