Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-(difluoromethyl)-5-fluorobenzene 97%

Supplier: AMBEED, INC
Warning

A733632-100MG A733632-1G A733632-250MG A733632-5G
76831-792EA 12 USD InStock
76831-792 76831-794 76831-796 76831-798
1-Bromo-3-(difluoromethyl)-5-fluorobenzene 97%
1-Bromo-3-(difluoromethyl)-5-fluorobenzene
Precaución: for Research Use Only


Formula: C₇H₄BrF₃
MDL Number: MFCD09261145
CAS Number: 627526-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty