Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di(pyridin-4-yl)methanone 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A735816-100MG A735816-250MG A735816-1G
76831-816EA 80 USD InStock
76831-816 76831-818 77684-659
Di(pyridin-4-yl)methanone 95%, Ambeed.Inc
Di(pyridin-4-yl)methanone
Precaución: For research use only.


Formula: C₁₁H₈N₂O
MDL Number: MFCD00832238
CAS Number: 6918-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty