Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Tosyl-7H-Pyrrolo[2,3-d]pyrimidine-5-boronic acid pinacol ester 98+%

Supplier: AMBEED, INC
Warning

A737822-100MG A737822-1G A737822-250MG A737822-5G
76831-830EA 36 USD InStock
76831-830 76831-832 76831-834 76831-836
7-Tosyl-7H-Pyrrolo[2,3-d]pyrimidine-5-boronic acid pinacol ester 98+%
7-Tosyl-7H-Pyrrolo[2,3-d]pyrimidine-5-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₉H₂₂BN₃O₄S
MDL Number: MFCD17214383
CAS Number: 934178-97-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty