Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-iodo-5-nitro-1H-indole-1-carboxylate 95%

Supplier: AMBEED, INC

A747224-100MG A747224-1G A747224-250MG A747224-5G
76831-916EA 5 USD InStock
76831-916 76831-918 76831-920 76831-922
tert-Butyl 3-iodo-5-nitro-1H-indole-1-carboxylate 95%
tert-Butyl 3-iodo-5-nitro-1H-indole-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₃IN₂O₄
MDL Number: MFCD05864777
CAS Number: 914349-32-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty