Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BMS-777607 98%

Supplier: AMBEED, INC
Warning

A756664-1MG A756664-5MG A756664-10MG
76831-986EA 18 USD InStock
76831-986 76831-988 77614-336
BMS-777607 98%
BMS-777607
Precaución: for Research Use Only


Formula: C₂₅H₁₉ClF₂N₄O₄
MW: 512.89 g/mol
MDL Number: MFCD16495773
CAS Number: 1025720-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty