Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Cyclopropylmethoxy)-5-pyridineboronic acid 98%

Supplier: AMBEED, INC
Warning

A759077-100MG A759077-1G A759077-250MG
76831-998EA 12 USD InStock
76831-998 76832-000 76832-002
2-(Cyclopropylmethoxy)-5-pyridineboronic acid 98%
2-(Cyclopropylmethoxy)-5-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₂BNO₃
MW: 193.0100 g/mol
MDL Number: MFCD12964560
CAS Number: 1028749-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty