Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (5-nitropyridin-2-yl)carbamate 95%

Supplier: AMBEED, INC
Warning

A774541-1G A774541-250MG A774541-5G
76832-080EA 42 USD InStock
76832-080 76832-082 76832-084
tert-Butyl (5-nitropyridin-2-yl)carbamate 95%
tert-Butyl (5-nitropyridin-2-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₃N₃O₄
MDL Number: MFCD11977408
CAS Number: 161117-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty