Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyltriphenylphosphonium bromide 98%

Supplier: AMBEED, INC
Warning

A792808-100G A792808-25G A792808-5G
76832-252EA 122 USD InStock
76832-252 76832-254 76832-256
Isopropyltriphenylphosphonium bromide 98%
Isopropyltriphenylphosphonium bromide
Precaución: for Research Use Only


Formula: C₂₁H₂₂BrP
MDL Number: MFCD00064807
CAS Number: 1530-33-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty