Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Methyl-1-naphthaldehyde 95%

Supplier: AMBEED, INC
Warning

A796396-100MG A796396-1G A796396-250MG
76832-286EA 35 USD InStock
76832-286 76832-288 76832-290
8-Methyl-1-naphthaldehyde 95%
8-Methyl-1-naphthaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₁₀O
MW: 170.20 g/mol
MDL Number: MFCD16293732
CAS Number: 6549-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty