Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-3-pyridineboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Danger

A801156-1G A801156-250MG A801156-5G
76832-334EA 72 USD InStock
76832-334 76832-336 76832-338
2,5-Dichloro-3-pyridineboronic acid pinacol ester 98%
2,5-Dichloro-3-pyridineboronic acid pinacol ester


Formula: C₁₁H₁₄BCl₂NO₂
MDL Number: MFCD08063094
CAS Number: 1073371-98-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty