Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-cyano-3-(trifluoromethyl)benzoate 95%

Supplier: AMBEED, INC
Warning

A810151-100MG A810151-250MG A810151-1G
76832-370EA 30 USD InStock
76832-370 76832-372 77308-368
Methyl 2-cyano-3-(trifluoromethyl)benzoate 95%
Methyl 2-cyano-3-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₆F₃NO₂
MDL Number: MFCD11109765
CAS Number: 1211596-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty