Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanfacine hydrochloride 99%

Supplier: AMBEED, INC
Danger

A810155-100MG A810155-25MG A810155-50MG A810155-1G
76832-374EA 11 USD InStock
76832-374 76832-376 76832-378 77614-444
Guanfacine hydrochloride 99%
Guanfacine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₉Cl₂N₃O
MW: 282.55 g/mol
MDL Number: MFCD00798230
CAS Number: 29110-48-3
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty