Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,5-Dichlorophenyl)-2,2,2-trifluoroethanone 95%

Supplier: AMBEED, INC
Warning

A812041-100MG A812041-1G A812041-250MG
76832-390EA 26 USD InStock
76832-390 76832-392 76832-394
1-(2,5-Dichlorophenyl)-2,2,2-trifluoroethanone 95%
1-(2,5-Dichlorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only


Formula: C₈H₃Cl₂F₃O
MDL Number: MFCD02260845
CAS Number: 886371-22-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty