Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis[(S)-4-isopropyl-4,5-dihydrooxazol-2-yl]methane 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A819829-100MG A819829-250MG A819829-50MG A819829-1G
76832-404EA 14 USD InStock
76832-404 76832-406 76832-408 77684-775
Bis[(S)-4-isopropyl-4,5-dihydrooxazol-2-yl]methane 97%, Ambeed.Inc
Bis[(S)-4-isopropyl-4,5-dihydrooxazol-2-yl]methane
Precaución: For research use only.


Formula: C₁₃H₂₂N₂O₂
MDL Number: MFCD11558948
CAS Number: 131833-90-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty