Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PD 184,352 98%

Supplier: AMBEED, INC
Warning

A832312-100MG A832312-10MG A832312-1G A832312-1MG A832312-250MG A832312-25MG A832312-50MG A832312-5MG
76832-492EA 267 USD InStock
76832-492 76832-494 76832-496 76832-498 76832-500 76832-502 76832-504 76832-506
PD 184,352 98%
PD 184,352
Precaución: for Research Use Only


Formula: C₁₇H₁₄ClF₂IN₂O₂
MW: 478.7 g/mol
MDL Number: MFCD02683961
CAS Number: 212631-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty