Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A838799-1G A838799-250MG A838799-5G
76832-530EA 22 USD InStock
76832-530 76832-532 76832-534
2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester 98%
2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₅BF₃NO₂
MW: 297.0806 g/mol
MDL Number: MFCD09878539
CAS Number: 1073355-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty