Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(4-Methoxybenzyl)sulfamoyl]benzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A859991-1G A859991-250MG A859991-5G
76832-612EA 94 USD InStock
76832-612 76832-614 76832-616
4-[(4-Methoxybenzyl)sulfamoyl]benzeneboronic acid 98%
4-[(4-Methoxybenzyl)sulfamoyl]benzeneboronic acid


Formula: C₁₄H₁₆BNO₅S
MDL Number: MFCD09027228
CAS Number: 957060-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty