Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

rel-(5s,6R,8r,9aS)-3-Oxooctahydro-1H-2,6-methanoquinolizin-8-yl 1H-indole-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A868963-10MG A868963-1MG A868963-5MG
76832-642EA 29 USD InStock
76832-642 76832-644 76832-646
rel-(5s,6R,8r,9aS)-3-Oxooctahydro-1H-2,6-methanoquinolizin-8-yl 1H-indole-3-carboxylate 98%
rel-(5s,6R,8r,9aS)-3-Oxooctahydro-1H-2,6-methanoquinolizin-8-yl 1H-indole-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₉H₂₀N₂O₃
MDL Number: MFCD00865542
CAS Number: 115956-12-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty