Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzylamino)acetonitrile 95%

Supplier: AMBEED, INC
Danger

A873760-100MG A873760-1G A873760-250MG A873760-5G
76832-688EA 11 USD InStock
76832-688 76832-690 76832-692 76832-694
2-(Benzylamino)acetonitrile 95%
2-(Benzylamino)acetonitrile
Precaución: for Research Use Only


Formula: C₉H₁₀N₂
MDL Number: MFCD00597541
CAS Number: 3010-05-7
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty