Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)phenethyl decanoate 95%

Supplier: AMBEED, INC
Warning

A898987-100G A898987-25G A898987-5G
76832-846EA 91 USD InStock
76832-846 76832-848 76832-850
4-(Benzyloxy)phenethyl decanoate 95%
4-(Benzyloxy)phenethyl decanoate
Precaución: for Research Use Only


Formula: C₂₅H₃₄O₃
MDL Number: MFCD22493501
CAS Number: 848484-93-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty