Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-1H-indole-2-carboxylic acid methyl ester 98%

Supplier: AMBEED, INC
Warning

A918281-100MG A918281-1G A918281-250MG
76832-904EA 39 USD InStock
76832-904 76832-906 76832-908
4-Methyl-1H-indole-2-carboxylic acid methyl ester 98%
4-Methyl-1H-indole-2-carboxylic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MDL Number: MFCD04966966
CAS Number: 136831-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty