Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-indoleboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A928830-100MG A928830-1G A928830-250MG
76832-938EA 28 USD InStock
76832-938 76832-940 76832-942
5-Chloro-2-indoleboronic acid pinacol ester 98%
5-Chloro-2-indoleboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₇BClNO₂
MDL Number: MFCD11858358
CAS Number: 1256358-91-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty