Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(4-phenoxyphenyl)acetate 97+%

Supplier: AMBEED, INC
Warning

A952972-1G A952972-250MG A952972-5G
76833-040EA 40 USD InStock
76833-040 76833-042 76833-044
Ethyl 2-(4-phenoxyphenyl)acetate 97+%
Ethyl 2-(4-phenoxyphenyl)acetate
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₃
MDL Number: MFCD12828046
CAS Number: 14062-26-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty