Virus Recombinant SARS-CoV-2 Omicron BA.4/5 Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Supplier: REVACC INC
B90Y94-100UG B90Y94-500UG
76729-692EA 383.91 USD InStock
76729-692 76729-552
Virus Recombinant SARS-CoV-2 Omicron BA.4/5 Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Proteins and Peptides
A recombinant form of the receptor binding domain (RBD) with amino acids 319 to 541) of the spike (S) glycoprotein gene from SARS-CoV-2 bearing mutations identified in variant of Omicron BA.4/5, was produced by insect cells, followed by purification.
  • This product contains C-terminal 6 hexa-histidine tag
This product has been run on SDS-PAGE with reduced condition. We also evaluated its binding to ACE2-Fc, anti-His mAb and a negative mAb in ELISAs.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty