Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylpyridine 98%

Supplier: AMBEED, INC
Warning

A284316-25G A284316-1000G A284316-100G A284316-10G A284316-500G A284316-5G
76837-298EA 8 USD InStock
76837-298 77289-714 77289-716 77289-718 77289-720 77289-722
4-Acetylpyridine 98%
4-Acetylpyridine
Precaución: for Research Use Only


Formula: C₇H₇NO
MDL Number: MFCD00006433
CAS Number: 1122-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty