Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Isopropyl-2-methylphenol 98%

Supplier: AMBEED, INC
Warning

A288147-25G A288147-1000G A288147-100G A288147-100MG A288147-500G
76837-478EA 10 USD InStock
76837-478 76977-530 77568-586 77571-784 77672-482
5-Isopropyl-2-methylphenol 98%
5-Isopropyl-2-methylphenol
Precaución: For research use only.


Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237
Melting Pt: 0.5
MDL Number: MFCD00002236
CAS Number: 499-75-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty